Learn More
Medchemexpress LLC K-Ras ligand-Linker Conjugate 4 | 2378261-83-5 | 98.8% | 25 MG
SDP
Catalog No. 5000341426
Encompass_Preferred
K-Ras ligand-Linker Conjugate 4 incorporates a ligand for the K-Ras recruiting moiety and a PROTAC linker. This linker recruits E3 ligases such as VHL, CRBN, MDM2, and IAP. It can be used in the synthesis of PROTAC K-Ras Degrader-1, a potent PROTAC K-Ras degrader that achieves ≥70% degradation efficacy in SW1573 cells. PROTACs consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates a ligand for the K-Ras recruiting moiety
  • Includes a PROTAC linker that recruits E3 ligases (VHL, CRBN, MDM2, and IAP)
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Can be used in the synthesis of PROTAC K-Ras Degrader-1
  • For research use only
Catalog No. 50-003-41426 Supplier Medchemexpress LLC Supplier No. 25MGHY130822
K-Ras ligand-Linker Conjugate 4 incorporates a ligand for the K-Ras recruiting moiety and a PROTAC linker. This linker recruits E3 ligases such as VHL, CRBN, MDM2, and IAP. It can be used in the synthesis of PROTAC K-Ras Degrader-1, a potent PROTAC K-Ras degrader that achieves ≥70% degradation efficacy in SW1573 cells. PROTACs consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates a ligand for the K-Ras recruiting moiety
  • Includes a PROTAC linker that recruits E3 ligases (VHL, CRBN, MDM2, and IAP)
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Can be used in the synthesis of PROTAC K-Ras Degrader-1
  • For research use only