Learn More
Medchemexpress LLC K-Ras ligand-Linker Conjugate 4 | 2378261-83-5 | 98.8% | 50 MG
SDP
Catalog No. 5000340535
Encompass_Preferred
K-Ras ligand-Linker Conjugate 4 integrates a ligand for K-Ras recruiting moiety and a PROTAC linker, which recruits E3 ligases (VHL, CRBN, MDM2, IAP). It is used in the synthesis of a potent PROTAC K-Ras degrader that demonstrates ≥70% degradation efficacy in SW1573 cells. PROTACs utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • Incorporates a ligand for K-Ras recruiting moiety.
  • Features a PROTAC linker to recruit E3 ligases.
  • Used in the synthesis of potent protein degraders.
  • Facilitates selective degradation of target proteins.
  • Leverages the ubiquitin-proteasome system.
Catalog No. 50-003-40535 Supplier Medchemexpress LLC Supplier No. HY13082250MG
K-Ras ligand-Linker Conjugate 4 integrates a ligand for K-Ras recruiting moiety and a PROTAC linker, which recruits E3 ligases (VHL, CRBN, MDM2, IAP). It is used in the synthesis of a potent PROTAC K-Ras degrader that demonstrates ≥70% degradation efficacy in SW1573 cells. PROTACs utilize the intracellular ubiquitin-proteasome system for selective degradation of target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • Incorporates a ligand for K-Ras recruiting moiety.
  • Features a PROTAC linker to recruit E3 ligases.
  • Used in the synthesis of potent protein degraders.
  • Facilitates selective degradation of target proteins.
  • Leverages the ubiquitin-proteasome system.