Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras ligand-linker conjugate 4 | 2378261-83-5 | 98.8% | 697.82 | C35H51N7O8 | 5 MG
SDP

Catalog No. 5000262137
Encompass_Preferred

K-Ras ligand-Linker Conjugate 4 is a PROTAC-style ligand-linker conjugate that combines a KRAS-targeting ligand with a PROTAC linker for use in degrader synthesis and related biochemical research. It is supplied as an oily, colorless to light-yellow material intended for laboratory research applications.

  • High purity: 98.8% by HPLC.
  • Molecular weight: 697.82 g/mol.
  • Appearance: oily, colorless to light yellow.
  • Storage: store at -20°C under nitrogen; in solvent store at -80°C up to 6 months.
  • Available quantities: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, larger sizes on request.
  • Analytical data: COA, HNMR, LC-MS, and RP-HPLC available.
  • Intended use: for synthesis of PROTACs and biochemical research only.

Catalog No. 50-002-62137 Supplier Medchemexpress LLC Supplier No. HY1308225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras ligand-Linker Conjugate 4 is a PROTAC-style ligand-linker conjugate that combines a KRAS-targeting ligand with a PROTAC linker for use in degrader synthesis and related biochemical research. It is supplied as an oily, colorless to light-yellow material intended for laboratory research applications.

  • High purity: 98.8% by HPLC.
  • Molecular weight: 697.82 g/mol.
  • Appearance: oily, colorless to light yellow.
  • Storage: store at -20°C under nitrogen; in solvent store at -80°C up to 6 months.
  • Available quantities: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, larger sizes on request.
  • Analytical data: COA, HNMR, LC-MS, and RP-HPLC available.
  • Intended use: for synthesis of PROTACs and biochemical research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.