Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras ligand-linker conjugate 4 | 2378261-83-5 | 98.8% | C35H51N7O8 | 10MG
SDP

Catalog No. 5000259874
Encompass_Preferred

K-Ras ligand-Linker Conjugate 4 is a research-grade ligand-linker conjugate designed for K-Ras targeting and PROTAC development. Supplied as an oil or as a 10 mM solution in ethanol, it is intended for laboratory research use only.

  • High purity (98.76%).
  • Available in multiple pack sizes, including 10 mg and solution format.
  • Appearance: oil; soluble in ethanol for solution preparations.
  • Storage: -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
  • Suitable for biochemical and chemical biology research involving ligand conjugation and PROTAC studies.

Catalog No. 50-002-59874 Supplier Medchemexpress LLC Supplier No. HY13082210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras ligand-Linker Conjugate 4 is a research-grade ligand-linker conjugate designed for K-Ras targeting and PROTAC development. Supplied as an oil or as a 10 mM solution in ethanol, it is intended for laboratory research use only.

  • High purity (98.76%).
  • Available in multiple pack sizes, including 10 mg and solution format.
  • Appearance: oil; soluble in ethanol for solution preparations.
  • Storage: -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
  • Suitable for biochemical and chemical biology research involving ligand conjugation and PROTAC studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.