Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-ras ligand-linker conjugate 5 | 2378261-85-7 | 99.8% | 10 MM 1 ML
SDP

Catalog No. 5000338542
Encompass_Preferred

K-Ras ligand-Linker Conjugate 5 is a chemical compound with a molecular formula of C42H59N7O8 and a molecular weight of 789.96. It boasts a high purity of 99.79% as confirmed by LCMS. This product is provided for research use only and has not been fully validated for medical applications.

  • Purity of 99.79% (LCMS)
  • Structure consistent with 1H NMR spectrum and LCMS
  • Intended for research use only
  • Molecular formula: C42H59N7O8
  • Molecular weight: 789.96

Catalog No. 50-003-38542 Supplier Medchemexpress LLC Supplier No. HY13082310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras ligand-Linker Conjugate 5 is a chemical compound with a molecular formula of C42H59N7O8 and a molecular weight of 789.96. It boasts a high purity of 99.79% as confirmed by LCMS. This product is provided for research use only and has not been fully validated for medical applications.

  • Purity of 99.79% (LCMS)
  • Structure consistent with 1H NMR spectrum and LCMS
  • Intended for research use only
  • Molecular formula: C42H59N7O8
  • Molecular weight: 789.96

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.