Learn More
Medchemexpress LLC K-Ras ligand-linker conjugate 5 | 2378261-85-7 | 99.8% | 789.96 g·mol⁻¹ | C42H59N7O8 | 5 MG
SDP
Catalog No. 5000263020
Encompass_Preferred
K-Ras ligand-linker conjugate 5 is a synthetic small-molecule conjugate that links a K-Ras-binding ligand to a PROTAC-style linker for use in targeted protein degradation research. It is intended as a building block for synthesis of K-Ras degraders and related cellular assay development, supplied in a small research-scale quantity.
  • High chemical purity suitable for research applications.
  • Suitable as a building block for proteolysis targeting chimera synthesis.
  • Defined molecular formula and molecular weight to support synthesis planning.
  • Provided in a research-scale quantity for assay development and pilot synthesis.
  • Compatible with standard organic synthesis and conjugation workflows.
Catalog No. 50-002-63020 Supplier Medchemexpress LLC Supplier No. HY1308235MG
K-Ras ligand-linker conjugate 5 is a synthetic small-molecule conjugate that links a K-Ras-binding ligand to a PROTAC-style linker for use in targeted protein degradation research. It is intended as a building block for synthesis of K-Ras degraders and related cellular assay development, supplied in a small research-scale quantity.
  • High chemical purity suitable for research applications.
  • Suitable as a building block for proteolysis targeting chimera synthesis.
  • Defined molecular formula and molecular weight to support synthesis planning.
  • Provided in a research-scale quantity for assay development and pilot synthesis.
  • Compatible with standard organic synthesis and conjugation workflows.