Learn More
Medchemexpress LLC K-Ras ligand-Linker Conjugate 5 | 97.0% | 50 MG
SDP
Catalog No. 5000339003
Encompass_Preferred
K-Ras ligand-Linker Conjugate 5 incorporates a ligand for K-Ras recruiting moiety, and a PROTAC linker, which recruit E3 ligases (such as VHL, CRBN, MDM2, and IAP).
  • Incorporates a ligand for K-Ras recruiting moiety
  • Features a PROTAC linker
  • Recruits E3 ligases (such as VHL, CRBN, MDM2, and IAP)
Catalog No. 50-003-39003 Supplier Medchemexpress LLC Supplier No. HY13082350MG
K-Ras ligand-Linker Conjugate 5 incorporates a ligand for K-Ras recruiting moiety, and a PROTAC linker, which recruit E3 ligases (such as VHL, CRBN, MDM2, and IAP).
  • Incorporates a ligand for K-Ras recruiting moiety
  • Features a PROTAC linker
  • Recruits E3 ligases (such as VHL, CRBN, MDM2, and IAP)