Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras ligand-linker conjugate 6 | 00-00-0 | C42H60N8O7 | 10MG
SDP

Catalog No. 5000258895
Encompass_Preferred

K-Ras ligand-linker conjugate 6 is a research-grade ligand-linker conjugate intended for K-Ras targeting and PROTAC development. Supplied as a solid for biochemical and cellular research, it has formula C42H60N8O7, molecular weight 788.98 g·mol⁻¹, and CAS 2378261-89-1. Store dry under inert gas at -20°C; in solvent store at -80°C for up to 6 months.

  • Designed for K-Ras targeting in PROTAC and ligand-linker studies.
  • Suitable for biochemical and cellular research applications.
  • Provided in multiple milligram-scale quantities for flexible dosing.
  • Characterized by molecular formula C42H60N8O7 and MW 788.98 g·mol⁻¹.
  • Includes datasheet and safety data sheet for handling and storage guidance.

Catalog No. 50-002-58895 Supplier Medchemexpress LLC Supplier No. HY130991A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras ligand-linker conjugate 6 is a research-grade ligand-linker conjugate intended for K-Ras targeting and PROTAC development. Supplied as a solid for biochemical and cellular research, it has formula C42H60N8O7, molecular weight 788.98 g·mol⁻¹, and CAS 2378261-89-1. Store dry under inert gas at -20°C; in solvent store at -80°C for up to 6 months.

  • Designed for K-Ras targeting in PROTAC and ligand-linker studies.
  • Suitable for biochemical and cellular research applications.
  • Provided in multiple milligram-scale quantities for flexible dosing.
  • Characterized by molecular formula C42H60N8O7 and MW 788.98 g·mol⁻¹.
  • Includes datasheet and safety data sheet for handling and storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.