Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras ligand-Linker Conjugate 6 | 2378261-89-1 | 100 MG
SDP

Catalog No. 5000340313
Encompass_Preferred

K-Ras ligand-Linker Conjugate 6 is a chemical compound with the formula C42H60N8O7 and a molecular weight of 788.98. This product appears as an oil and is stable under recommended storage conditions. It is intended for use as a laboratory chemical and for the manufacture of substances, strictly for research purposes by experienced personnel in appropriately equipped facilities.

  • Identified for laboratory chemical applications
  • Chemical formula: C42H60N8O7
  • Molecular weight: 788.98
  • Physical appearance: oil
  • Stable under recommended storage conditions
  • May cause skin and serious eye irritation
  • May cause respiratory irritation
  • Harmful if swallowed

Catalog No. 50-003-40313 Supplier Medchemexpress LLC Supplier No. HY130991A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras ligand-Linker Conjugate 6 is a chemical compound with the formula C42H60N8O7 and a molecular weight of 788.98. This product appears as an oil and is stable under recommended storage conditions. It is intended for use as a laboratory chemical and for the manufacture of substances, strictly for research purposes by experienced personnel in appropriately equipped facilities.

  • Identified for laboratory chemical applications
  • Chemical formula: C42H60N8O7
  • Molecular weight: 788.98
  • Physical appearance: oil
  • Stable under recommended storage conditions
  • May cause skin and serious eye irritation
  • May cause respiratory irritation
  • Harmful if swallowed

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.