Learn More
Medchemexpress LLC K-Ras ligand-linker conjugate 6 | 2378261-89-1 | 95.0% | 788.98 g/mol | C42H60N8O7 | 5 MG
SDP
Catalog No. 5000262161
Encompass_Preferred
K-Ras ligand-Linker Conjugate 6 is a ligand-linker conjugate designed as a synthetic building block for assembling PROTAC degraders that target KRAS. It combines a KRAS-binding ligand with a PROTAC-compatible linker to enable recruitment of E3 ligases for targeted protein degradation and is supplied as a small-mass research reagent for medicinal chemistry and degrader design.
  • Facilitates assembly of KRAS-directed PROTAC molecules.
  • Enables recruitment of common E3 ligases such as VHL, CRBN, MDM2, and IAP.
  • Provided in small-mass quantities suitable for reaction screening and optimization.
  • Compatible with standard synthetic coupling chemistries for conjugation.
  • Characterized with reported molecular formula and molecular weight for tracking.
Catalog No. 50-002-62161 Supplier Medchemexpress LLC Supplier No. HY1309915MG
K-Ras ligand-Linker Conjugate 6 is a ligand-linker conjugate designed as a synthetic building block for assembling PROTAC degraders that target KRAS. It combines a KRAS-binding ligand with a PROTAC-compatible linker to enable recruitment of E3 ligases for targeted protein degradation and is supplied as a small-mass research reagent for medicinal chemistry and degrader design.
  • Facilitates assembly of KRAS-directed PROTAC molecules.
  • Enables recruitment of common E3 ligases such as VHL, CRBN, MDM2, and IAP.
  • Provided in small-mass quantities suitable for reaction screening and optimization.
  • Compatible with standard synthetic coupling chemistries for conjugation.
  • Characterized with reported molecular formula and molecular weight for tracking.