Learn More
Medchemexpress LLC K-Ras ligand-linker conjugate 6 | 2378261-89-1 | C42H60N8O7 | 10MG
SDP
Catalog No. 5000259884
Encompass_Preferred
K-Ras ligand-Linker Conjugate 6 is a small-molecule ligand-linker designed as an intermediate for targeted protein degradation research. It links a K-Ras-binding ligand to a PROTAC linker to enable synthesis of K-Ras degraders for cellular degradation studies.
  • High purity confirmed by HPLC (98.9%).
  • Characterized by mass spectrometry consistent with structure.
  • Molecular formula C42H60N8O7 and molecular weight 788.98 g/mol.
  • Suitable for PROTAC synthesis and K-Ras degrader development.
  • Available in multiple small-scale pack sizes (5 mg-100 mg) for research use.
Catalog No. 50-002-59884 Supplier Medchemexpress LLC Supplier No. HY13099110MG
K-Ras ligand-Linker Conjugate 6 is a small-molecule ligand-linker designed as an intermediate for targeted protein degradation research. It links a K-Ras-binding ligand to a PROTAC linker to enable synthesis of K-Ras degraders for cellular degradation studies.
  • High purity confirmed by HPLC (98.9%).
  • Characterized by mass spectrometry consistent with structure.
  • Molecular formula C42H60N8O7 and molecular weight 788.98 g/mol.
  • Suitable for PROTAC synthesis and K-Ras degrader development.
  • Available in multiple small-scale pack sizes (5 mg-100 mg) for research use.