Learn More
Medchemexpress LLC K-Ras ligand-Linker Conjugate 6 formate | 98.9% | 25 MG
SDP
Catalog No. 5000341695
Encompass_Preferred
K-Ras ligand-Linker Conjugate 6 formate is a chemical compound with the molecular formula C43H62N8O9 and a molecular weight of 835.00. This compound is provided in an off-white to light yellow solid form and is suitable for various laboratory applications requiring high purity chemical reagents.
  • High purity of 98.93% verified by HPLC
  • Appears as an off-white to light yellow solid
  • Store at -20°C in sealed conditions, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month in sealed conditions
  • Analytical data (1H NMR spectrum, MS) are consistent with its structure
Catalog No. 50-003-41695 Supplier Medchemexpress LLC Supplier No. 25MGHY130991A
K-Ras ligand-Linker Conjugate 6 formate is a chemical compound with the molecular formula C43H62N8O9 and a molecular weight of 835.00. This compound is provided in an off-white to light yellow solid form and is suitable for various laboratory applications requiring high purity chemical reagents.
  • High purity of 98.93% verified by HPLC
  • Appears as an off-white to light yellow solid
  • Store at -20°C in sealed conditions, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month in sealed conditions
  • Analytical data (1H NMR spectrum, MS) are consistent with its structure