Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras-PDE -IN-1 100mg | 1841464-21-8 | 447.50 g·mol⁻1 | C25H26FN5O2 | 100 MG
SDP

Catalog No. 5000226688
Encompass_Preferred

K-Ras-PDEδ-IN-1 is a small-molecule, competitive inhibitor of PDEδ that binds the farnesyl-binding pocket with low-nanomolar affinity (Kd = 8 nM). It is supplied as a high-purity research chemical suitable for biochemical and cellular studies, with solubility and storage data provided for experimental use.

  • Potent PDEδ inhibitor with a reported Kd of 8 nM.
  • Binds the farnesyl-binding pocket of PDEδ.
  • High purity (99.62%) suitable for research applications.
  • Molecular formula C25H26FN5O2 and molecular weight 447.50 g·mol⁻1.
  • Soluble in DMSO (12.5 mg/mL; sonication recommended).
  • Stable as powder under low-temperature storage (powder: -20°C for 3 years; 4°C for 2 years).
  • Available in a 100 MG pack size for laboratory use.

Catalog No. 50-002-26688 Supplier Medchemexpress LLC Supplier No. HY115555100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras-PDEδ-IN-1 is a small-molecule, competitive inhibitor of PDEδ that binds the farnesyl-binding pocket with low-nanomolar affinity (Kd = 8 nM). It is supplied as a high-purity research chemical suitable for biochemical and cellular studies, with solubility and storage data provided for experimental use.

  • Potent PDEδ inhibitor with a reported Kd of 8 nM.
  • Binds the farnesyl-binding pocket of PDEδ.
  • High purity (99.62%) suitable for research applications.
  • Molecular formula C25H26FN5O2 and molecular weight 447.50 g·mol⁻1.
  • Soluble in DMSO (12.5 mg/mL; sonication recommended).
  • Stable as powder under low-temperature storage (powder: -20°C for 3 years; 4°C for 2 years).
  • Available in a 100 MG pack size for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.