Learn More
Medchemexpress LLC K-Ras-PDE -IN-1 | 10MG
SDP
Catalog No. 5000227918
Encompass_Preferred
K-Ras-PDE -IN-1 is a novel and potent competitive K-Ras-PDE inhibitor K-Ras-PDE -IN-1 binds to the farnesyl binding pocket of PDE with a low nanomolar Kd of 8 nM[1]
Catalog No. 50-002-27918 Supplier Medchemexpress LLC Supplier No. HY11555510MG
K-Ras-PDE -IN-1 is a novel and potent competitive K-Ras-PDE inhibitor K-Ras-PDE -IN-1 binds to the farnesyl binding pocket of PDE with a low nanomolar Kd of 8 nM[1]