Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras-PDE -IN-1 25mg | 1841464-21-8 | 447.50 g/mol | C25H26FN5O2 | 25 MG
SDP

Catalog No. 5000227761
Encompass_Preferred

K-Ras-PDEδ-IN-1 is a small-molecule research compound that competitively inhibits PDEδ by binding the farnesyl-binding pocket with low nanomolar affinity (Kd ≈ 8 nM). It is supplied as a solid for in vitro and in vivo research applications and includes solubility and formulation guidance for biomedical studies.

  • Potent competitive inhibitor of PDEδ with Kd ≈ 8 nM.
  • Intended for use in Ras/PDEδ biological research in vitro and in vivo.
  • Soluble in DMSO at about 12.5 mg/mL; in vivo suspension guidance at ~1.25 mg/mL with standard vehicle.
  • Molecular formula C25H26FN5O2 and molecular weight 447.50 g/mol.
  • Supplied as a solid powder suitable for biochemical and cellular assays.

Catalog No. 50-002-27761 Supplier Medchemexpress LLC Supplier No. HY11555525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras-PDEδ-IN-1 is a small-molecule research compound that competitively inhibits PDEδ by binding the farnesyl-binding pocket with low nanomolar affinity (Kd ≈ 8 nM). It is supplied as a solid for in vitro and in vivo research applications and includes solubility and formulation guidance for biomedical studies.

  • Potent competitive inhibitor of PDEδ with Kd ≈ 8 nM.
  • Intended for use in Ras/PDEδ biological research in vitro and in vivo.
  • Soluble in DMSO at about 12.5 mg/mL; in vivo suspension guidance at ~1.25 mg/mL with standard vehicle.
  • Molecular formula C25H26FN5O2 and molecular weight 447.50 g/mol.
  • Supplied as a solid powder suitable for biochemical and cellular assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.