Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras-PDE -IN-1 50mg | 1841464-21-8 | 447.50 g/mol | C25H26FN5O2 | 50 MG
SDP

Catalog No. 5000226719
Encompass_Preferred

K-Ras-PDEδ-IN-1 is a small-molecule research inhibitor that disrupts K-Ras-PDEδ interactions by binding the farnesyl binding pocket of PDEδ, with a reported Kd of 8 nM. It is supplied as a high-purity research reagent in solid and DMSO solution formats for in vitro and in vivo studies.

  • Selective inhibitor of K-Ras-PDEδ interaction (Kd 8 nM).
  • Binds the farnesyl binding pocket of PDEδ.
  • High purity: 99.62%.
  • Molecular formula C25H26FN5O2; molecular weight 447.50 g/mol.
  • Solubility: DMSO 12.5 mg/mL (in vitro); in vivo formulation protocol available.
  • Available as solids (5 mg-100 mg) and 10 mM DMSO solutions.

Catalog No. 50-002-26719 Supplier Medchemexpress LLC Supplier No. HY11555550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras-PDEδ-IN-1 is a small-molecule research inhibitor that disrupts K-Ras-PDEδ interactions by binding the farnesyl binding pocket of PDEδ, with a reported Kd of 8 nM. It is supplied as a high-purity research reagent in solid and DMSO solution formats for in vitro and in vivo studies.

  • Selective inhibitor of K-Ras-PDEδ interaction (Kd 8 nM).
  • Binds the farnesyl binding pocket of PDEδ.
  • High purity: 99.62%.
  • Molecular formula C25H26FN5O2; molecular weight 447.50 g/mol.
  • Solubility: DMSO 12.5 mg/mL (in vitro); in vivo formulation protocol available.
  • Available as solids (5 mg-100 mg) and 10 mM DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.