Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K-Ras-PDE -IN-1 5mg | 1841464-21-8 | 447.50 g/mol | C25H26FN5O2 | 5 MG
SDP

Catalog No. 5000307942
Encompass_Preferred

K-Ras-PDEδ-IN-1 is a small-molecule competitive inhibitor that binds the farnesyl binding pocket of PDEδ and disrupts PDEδ-K-Ras interaction. It is provided for research use in biochemical and cellular studies and is characterized by nanomolar affinity and high chemical purity. Follow supplier solubility and handling guidance when preparing solutions for in vitro or in vivo use.

  • Binds the PDEδ farnesyl pocket with a Kd of ~8 nM.
  • High chemical purity (≈99.6% as supplied).
  • Soluble in DMSO (12.5 mg/mL); ultrasonic recommended for dissolution.
  • Formulation guidance provided for in vivo suspension (10% DMSO, 40% PEG300, 5% Tween-80, 45% saline).
  • Available in small research pack sizes suitable for biochemical assays.

Catalog No. 50-003-07942 Supplier Medchemexpress LLC Supplier No. HY1155555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K-Ras-PDEδ-IN-1 is a small-molecule competitive inhibitor that binds the farnesyl binding pocket of PDEδ and disrupts PDEδ-K-Ras interaction. It is provided for research use in biochemical and cellular studies and is characterized by nanomolar affinity and high chemical purity. Follow supplier solubility and handling guidance when preparing solutions for in vitro or in vivo use.

  • Binds the PDEδ farnesyl pocket with a Kd of ~8 nM.
  • High chemical purity (≈99.6% as supplied).
  • Soluble in DMSO (12.5 mg/mL); ultrasonic recommended for dissolution.
  • Formulation guidance provided for in vivo suspension (10% DMSO, 40% PEG300, 5% Tween-80, 45% saline).
  • Available in small research pack sizes suitable for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.