Learn More
TARGETMOL CHEMICALS INC K-RASG12C INHIBITOR 12 5MG
SDP
Catalog No. 502576493
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. K-Ras(G12C) inhibitor 12 is an allosteric inhibitor of oncogenic K-Ras(G12C). purity: 99%
Catalog No. 50-257-6493 Supplier TARGETMOL CHEMICALS INC Supplier No. T65555MG
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. K-Ras(G12C) inhibitor 12 is an allosteric inhibitor of oncogenic K-Ras(G12C). purity: 99%