Learn More
Omega Engineering, Inc. K THERMOCOUPLE 24" 0-200C
Catalog No. NC2298750
Encompass
PFA Coated Thermocouple Probes for Chemical Environments. K type, .125" diameter probe, 0 to 200c, 24" length
Catalog No. NC2298750 Supplier Omega Engineering, Inc. Supplier No. CASS18G24PFA
PFA Coated Thermocouple Probes for Chemical Environments. K type, .125" diameter probe, 0 to 200c, 24" length