Learn More
Omega Engineering, Inc. K THERMOCOUPLE PROBE, 1/8 INCH
Catalog No. NC1590397
Encompass
K thermocouple probe, 1/8 inch diameter, 300mm(12 inch) length, grounded junction, molded connector
Catalog No. NC1590397 Supplier Omega Engineering, Inc. Supplier No. KMQXL125G12
K thermocouple probe, 1/8 inch diameter, 300mm(12 inch) length, grounded junction, molded connector