Learn More
Med Vet International K-Wire Bender Bender, J0211, Each
Catalog No. 5000585846
Encompass_Preferred
A unique tool for bending over the ends of K-wire after various osteotomies. Allows just the very tip to be bent over quickly and precisely. Works with all three sizes of K-wire.
Catalog No. 50-005-85846 Supplier Med Vet International Supplier No. JOR211
A unique tool for bending over the ends of K-wire after various osteotomies. Allows just the very tip to be bent over quickly and precisely. Works with all three sizes of K-wire.