Learn More
Med Vet International K-Wire Cutter, 8.5", J0220BA, Each
Catalog No. 5000585389
Encompass_Preferred
K-Wire End Cutter with tungsten carbide inserts. Cuts wire up to 2.5mm. 81/2"L.
Catalog No. 50-005-85389 Supplier Med Vet International Supplier No. JOR220BA
K-Wire End Cutter with tungsten carbide inserts. Cuts wire up to 2.5mm. 81/2"L.