Learn More
Sklar Instruments Vickers K-Wire Dispenser, Sklar, 13 in. for Wire 1mm, 1.2mm, 1.4mm, 1.6mm, Small, Sklar's Vickers K-Wire Dispenser is used to store and dispense four different size wires. Dial enables for dispensing of one size at a time. EA/1.
SDP
Catalog No. 501438333 Shop All Sklar Instruments Products
Encompass_Preferred
Vickers K-Wire Dispenser, Sklar, 13 in. for Wire 1mm, 1.2mm, 1.4mm, 1.6mm, Small, Sklar's Vickers K-Wire Dispenser is used to store and dispense four different size wires. Dial enables for dispensing of one size at a time. This product can lock to avoid acidental dispensing to avoid mix-up. The small dispenser has four chambers. Chamber 1 holds sixty pieces of 0.039 in. (1.0mm) wire; Chamber 2 holds forty pieces of 0.047 in. (1.2mm) wire; Chamber 3 holds thirty pieces of 0.055 in. (1.4mm) wire; Chamber 4 holds eighteen pieces of 0.063 in. (1.6mm) wire.
This item is not returnable. View return policy
Catalog No. 50-143-8333 Supplier Sklar Instruments Supplier No. 401576
Vickers K-Wire Dispenser, Sklar, 13 in. for Wire 1mm, 1.2mm, 1.4mm, 1.6mm, Small, Sklar's Vickers K-Wire Dispenser is used to store and dispense four different size wires. Dial enables for dispensing of one size at a time. This product can lock to avoid acidental dispensing to avoid mix-up. The small dispenser has four chambers. Chamber 1 holds sixty pieces of 0.039 in. (1.0mm) wire; Chamber 2 holds forty pieces of 0.047 in. (1.2mm) wire; Chamber 3 holds thirty pieces of 0.055 in. (1.4mm) wire; Chamber 4 holds eighteen pieces of 0.063 in. (1.6mm) wire.