Learn More
Cayman Chemical Ko 143, 461054-93-3, 5 mg
SDP
Supplier:  Cayman Chemical 152155
Encompass_Preferred
A potent and selective inhibitor of BCRP, preventing the export of mitoxantrone and topotecan in breast cancer cell lines (EC50s = 23 and 26 nM, respectively); effective in vivo in mice
Catalog No. NC1517135
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.