Learn More
TARGETMOL CHEMICALS INC K02288 25MG
SDP
Catalog No. 502581055
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K 02288 is a novel small molecule inhibitor of ALK1/2/3/6. purity: 99%
Catalog No. 50-258-1055 Supplier TARGETMOL CHEMICALS INC Supplier No. T191425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K 02288 is a novel small molecule inhibitor of ALK1/2/3/6. purity: 99%