Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC K1 peptide TFA | 98.2% | 1489.59 (free acid) | 1 MG
SDP

Catalog No. 5000397860
Encompass_Preferred

K1 peptide TFA is a high-affinity peptide ligand for GABAA receptor-associated protein (GABARAP).

  • High-affinity peptide ligand for GABAA receptor-associated protein (GABARAP)
  • Solid appearance
  • White to off-white color
  • Target: GABA Receptor
  • Pathway: Membrane Transporter/Ion Channel, Neuronal Signaling
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-97860 Supplier Medchemexpress LLC Supplier No. HYP5964A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K1 peptide TFA is a high-affinity peptide ligand for GABAA receptor-associated protein (GABARAP).

  • High-affinity peptide ligand for GABAA receptor-associated protein (GABARAP)
  • Solid appearance
  • White to off-white color
  • Target: GABA Receptor
  • Pathway: Membrane Transporter/Ion Channel, Neuronal Signaling
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.