Learn More
Enzo Life Sciences ([K11-only]Ub)n ubiquitinylated substrate (25 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW06200025
Encompass_Preferred
Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 150mM NaCl and 1mM DTT. Source: Protein components produced in E. coli. Synthesized enzymatically in vitro. (K11-only)Ubn-ubiquitinylated substrate protein was produced using K11-only mutant ubiquitin. Long Term Storage: -80°C.
Catalog No. 50-201-2256