Learn More
TARGETMOL CHEMICALS INC K145 HYDROCHLORIDE 50MG
SDP
Catalog No. 502576157
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg and bulk. Please contact Fisher for quotes. K145 hydrochloride is a selective sphk2 inhibitor with substrate competitiveness and oral activity with IC50 of 4.3 µM and Ki of 6.4 µM. K145 hydrochloride can induce apoptosis and has strong antitumor activity. purity: 99%
Catalog No. 50-257-6157 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ013850MG
Also available in 5 mg 10 mg 25 mg and bulk. Please contact Fisher for quotes. K145 hydrochloride is a selective sphk2 inhibitor with substrate competitiveness and oral activity with IC50 of 4.3 µM and Ki of 6.4 µM. K145 hydrochloride can induce apoptosis and has strong antitumor activity. purity: 99%