Learn More
Medchemexpress LLC HY-15779A 5mg Medchemexpress, K145 (hydrochloride) CAS:1449240-68-9 Purity:>98%
SDP
Catalog No. 502081798
Encompass_Preferred
Medchemexpress, HY-15779A 5mg K145 (hydrochloride) CAS:1449240-68-9 K145 hydrochloride is a selective, substrate-competitive and orally active SphK2 inhibitor with an IC50 of 4.3 µM and a Ki of 6.4 µM. K145 hydrochloride is inactive against SphK1 and other protein kinases. K145 hydrochloride induces cell apoptosis and has potently antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1798 Supplier Medchemexpress LLC Supplier No. HY15779A5MG
Medchemexpress, HY-15779A 5mg K145 (hydrochloride) CAS:1449240-68-9 K145 hydrochloride is a selective, substrate-competitive and orally active SphK2 inhibitor with an IC50 of 4.3 µM and a Ki of 6.4 µM. K145 hydrochloride is inactive against SphK1 and other protein kinases. K145 hydrochloride induces cell apoptosis and has potently antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.