Learn More
SAI Infusion Technologies Micro500 500uL Blood Tube- K2EDTA, Lavender Push Cap
SDP
Catalog No. NC0879116
Encompass
500ul Micro500 Blood Tube - K2EDTA, Lavender Cap, may be used for Plasma collection. SAI's Micro500 blood tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only.
Catalog No. NC0879116 Supplier SAI Infusion Technologies Supplier No. M500E
500ul Micro500 Blood Tube - K2EDTA, Lavender Cap, may be used for Plasma collection. SAI's Micro500 blood tubes replicate the dimensions of the discontinued BD Microtainer for 500ul blood collections. With the same slim 8 mm X 43 mm size, no adjustments for methods or instrumentation are required. 100/pack. Product is for laboratory animal research only.