Learn More
Electron Microscopy Sciences K20 Calibrated Cover Glass 1 ea
SDP
Catalog No. 5031232
Encompass_Preferred
The coverglass has 25 calibrated fields of 1.382mm diameter through which to view the particles. This coverglass removes the necessity of precise adjustment of the microscope and eyepiece graticule in the original Howard method.
Catalog No. 50-312-32 Supplier Electron Microscopy Sciences Supplier No. 6805120
The coverglass has 25 calibrated fields of 1.382mm diameter through which to view the particles. This coverglass removes the necessity of precise adjustment of the microscope and eyepiece graticule in the original Howard method.