Learn More
Abcam K252a, protein and CAM kinase inhibitor, 200UG
Catalog No. 502707603
Encompass_Preferred
MW 467.5 Da, Purity >99%. Highly potent, cell-permeable inhibitor of protein kinases such as PKA, PKC, PKG (IC₅₀ = 18-25 nM), Serine/threonine protein kinases (IC₅₀ values are 10-20 nM) and CAM kinase II (Ki = 1.8 nM). Analog of staurosporine (ab120056), exhibits a wide range of effects including neuroprotection and improvement of psoriasis in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7603 Supplier Abcam Supplier No. AB120419200UG
MW 467.5 Da, Purity >99%. Highly potent, cell-permeable inhibitor of protein kinases such as PKA, PKC, PKG (IC₅₀ = 18-25 nM), Serine/threonine protein kinases (IC₅₀ values are 10-20 nM) and CAM kinase II (Ki = 1.8 nM). Analog of staurosporine (ab120056), exhibits a wide range of effects including neuroprotection and improvement of psoriasis in vivo.
The product is subject to the following: Abcam Restricted Use Statement