Learn More
Grainger Ansell Ringers R-075 Impact Gloves, PVC Coated, Size 10, 1/Pk.
Catalog No. 502773708
Encompass_Preferred
The Ringers R-075 gauntlet-style gloves offer advanced chemical and mechanical protection for demanding work environments. Designed with F3 Technology and thermoplastic rubber, these gloves provide robust top-of-hand and full-finger impact protection. Engineered with a durable polyvinyl chloride coating and a specialized liner, they provide high-level resistance against chemical hazards and cuts, making them suitable for tasks such as drilling assembly and completion.
  • Provides finger-length and top-of-hand impact protection
  • Features a 360° cut-resistant protective liner
  • Meets EN 374-1 Type A chemical resistance standards (AJKLMO)
  • Constructed with a textured palm for enhanced grip
  • Compliant with ANSI/ISEA 105-2024 Cut Level A3 and EN ISO C standards
  • Certified for ANSI/ISEA 138 impact protection
  • Designed with a gauntlet cuff for extended forearm coverage
Catalog No. 50-277-3708 Supplier Grainger Supplier No. 181J29
The Ringers R-075 gauntlet-style gloves offer advanced chemical and mechanical protection for demanding work environments. Designed with F3 Technology and thermoplastic rubber, these gloves provide robust top-of-hand and full-finger impact protection. Engineered with a durable polyvinyl chloride coating and a specialized liner, they provide high-level resistance against chemical hazards and cuts, making them suitable for tasks such as drilling assembly and completion.
  • Provides finger-length and top-of-hand impact protection
  • Features a 360° cut-resistant protective liner
  • Meets EN 374-1 Type A chemical resistance standards (AJKLMO)
  • Constructed with a textured palm for enhanced grip
  • Compliant with ANSI/ISEA 105-2024 Cut Level A3 and EN ISO C standards
  • Certified for ANSI/ISEA 138 impact protection
  • Designed with a gauntlet cuff for extended forearm coverage