Learn More
Grainger Ansell Ringers R-074 Chemical Impact Gloves, PVC, Size 12, 1/Pk.
Catalog No. 502773706
Encompass_Preferred
These short gauntlet-style impact gloves provide durable, waterproof chemical resistance while ensuring enhanced safety and comfort. Designed with F3 Technology and thermoplastic rubber, these gloves offer robust impact protection across the top of the hand and full length of the fingers, making them suitable for demanding environments like automotive, manufacturing, and oil and gas.
  • Provides 360° cut resistance with an advanced inner liner
  • Features a sandy PVC textured palm for improved grip and durability
  • Tested against EN 374-1 Type A chemical resistance standards
  • Meets ANSI/ISEA 105-2024 Cut A3 and EN ISO C standards
  • Includes low-profile impact protection
  • Features a gauntlet cuff style for extended wrist protection
  • Non-sterile and chemical-resistant construction
Catalog No. 50-277-3706 Supplier Grainger Supplier No. 181J33
These short gauntlet-style impact gloves provide durable, waterproof chemical resistance while ensuring enhanced safety and comfort. Designed with F3 Technology and thermoplastic rubber, these gloves offer robust impact protection across the top of the hand and full length of the fingers, making them suitable for demanding environments like automotive, manufacturing, and oil and gas.
  • Provides 360° cut resistance with an advanced inner liner
  • Features a sandy PVC textured palm for improved grip and durability
  • Tested against EN 374-1 Type A chemical resistance standards
  • Meets ANSI/ISEA 105-2024 Cut A3 and EN ISO C standards
  • Includes low-profile impact protection
  • Features a gauntlet cuff style for extended wrist protection
  • Non-sterile and chemical-resistant construction