Learn More
SAI Infusion Technologies 1ml K2 EDTA Blood Collection Syringe with 25G5/8" needle
SDP
Catalog No. NC3432962
Encompass
1mL K2EDTA treated syringes with attached 25G 5/8" needles for blood collection. These syringes provide immediate anticoagulation, minimizing missed samples due to slow bleeds and clotting. 25/pack. Minimum order quantity of 12 packs required. Product is for laboratory animal research only.
Catalog No. NC3432962 Supplier SAI Infusion Technologies Supplier No. EDTAK2S1
1mL K2EDTA treated syringes with attached 25G 5/8" needles for blood collection. These syringes provide immediate anticoagulation, minimizing missed samples due to slow bleeds and clotting. 25/pack. Minimum order quantity of 12 packs required. Product is for laboratory animal research only.