Learn More
Grainger Fluke 9190A Field Metrology Well, -95 to 140°C, Process Electronics, 1 ea
Catalog No. 502795144
Encompass_Preferred
The Ultra-Cool Field Metrology Well is a highly accurate and stable cold temperature dry-block calibrator designed for on-location validation and calibration of RTDs, thermocouples, thermometers, and sensors. It is ideal for applications requiring strict quality control and regulatory process compliance, providing a cleanroom-compatible and portable solution that eliminates the need for heat transfer fluids.
  • Temperature range: -95°C to 140°C
  • Best-in-class accuracy of 0.05°C and stability of 0.015°C
  • Faster heating and cooling times compared to traditional calibration baths
  • Thoroughly tested specifications for axial and radial uniformity, hysteresis, and loading effects
  • Conforms to EURAMET cg-13 best practices for temperature block calibrators
  • Includes RS-232 and USB serial computer interfaces
Catalog No. 50-279-5144 Supplier Grainger Supplier No. 19YH33
Add to Cart
Add to Cart
Request a Quote
The Ultra-Cool Field Metrology Well is a highly accurate and stable cold temperature dry-block calibrator designed for on-location validation and calibration of RTDs, thermocouples, thermometers, and sensors. It is ideal for applications requiring strict quality control and regulatory process compliance, providing a cleanroom-compatible and portable solution that eliminates the need for heat transfer fluids.
  • Temperature range: -95°C to 140°C
  • Best-in-class accuracy of 0.05°C and stability of 0.015°C
  • Faster heating and cooling times compared to traditional calibration baths
  • Thoroughly tested specifications for axial and radial uniformity, hysteresis, and loading effects
  • Conforms to EURAMET cg-13 best practices for temperature block calibrators
  • Includes RS-232 and USB serial computer interfaces