Learn More
TARGETMOL CHEMICALS INC K34C HYDROCHLORIDE 5MG
SDP
Catalog No. 502579956
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K34c hydrochloride is an alpha5(beta)1 integrin antagonist for glioblastoma study.
Catalog No. 50-257-9956 Supplier TARGETMOL CHEMICALS INC Supplier No. T41151L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K34c hydrochloride is an alpha5(beta)1 integrin antagonist for glioblastoma study.