Learn More
Grainger K4796 BlueSiliconeRoundCord 1/16 D 100 L 70A
Catalog No. 502768148
Encompass_Preferred
The Silicone Round Cord Metal is a silicone cord with a metal core that is flexible and durable. It has a temperature range of -100°F to 500°F. The cord has a round cross section and comes in multiple diameters from 1/16 inch to 3/8 inch. It is made of platinum-cured silicone which resists weathering and aging. The metal core provides strength while allowing the cord to remain flexible. Common applications include gaskets, seals, hose sleeves, and more. The cord meets UL 94 V-0 flammability standards. It comes in standard 10 foot lengths.
Catalog No. 50-276-8148 Supplier Grainger Supplier No. 16Z140
Add to Cart
Add to Cart
The Silicone Round Cord Metal is a silicone cord with a metal core that is flexible and durable. It has a temperature range of -100°F to 500°F. The cord has a round cross section and comes in multiple diameters from 1/16 inch to 3/8 inch. It is made of platinum-cured silicone which resists weathering and aging. The metal core provides strength while allowing the cord to remain flexible. Common applications include gaskets, seals, hose sleeves, and more. The cord meets UL 94 V-0 flammability standards. It comes in standard 10 foot lengths.