Learn More
Grainger K4796 Silicone Round Cord 1/32 D 25 L 70A
Catalog No. 502767987
Encompass_Preferred
The Silicone Round Cord from Grainger is a silicone rubber cord that is round in shape. It is available in multiple diameters from 1/16 inch to 3/4 inch and comes in lengths of 25 feet. The cord has operating temperatures ranging from -80°F to 500°F and meets UL94 V-0 flammability standards. It is flexible and offers good resistance to weathering, moisture, and UV rays. The cord can be used for a variety of applications including weather seals, gaskets, vibration dampening, insulation, and more. It comes in red, clear or black colors. This silicone cord provides flexibility, durability, and high temperature resistance.
Catalog No. 50-276-7987 Supplier Grainger Supplier No. 16Z134
The Silicone Round Cord from Grainger is a silicone rubber cord that is round in shape. It is available in multiple diameters from 1/16 inch to 3/4 inch and comes in lengths of 25 feet. The cord has operating temperatures ranging from -80°F to 500°F and meets UL94 V-0 flammability standards. It is flexible and offers good resistance to weathering, moisture, and UV rays. The cord can be used for a variety of applications including weather seals, gaskets, vibration dampening, insulation, and more. It comes in red, clear or black colors. This silicone cord provides flexibility, durability, and high temperature resistance.