Learn More
Grainger K4796 Silicone Round Cord 1/8 D 25 L 70A Blue
Catalog No. 502767800
Encompass_Preferred
The Silicone Round Cord Metal is a silicone rubber cord with a metal core. It is made of high-quality silicone rubber that is flexible and durable. The metal core provides strength and shape retention. This cord has a round cross-section and comes in multiple diameters from 1/8" to 3/4". It can withstand temperatures from -80°F to 500°F. Potential applications include gaskets, seals, vibration dampening, noise reduction, and more. This product offers flexibility, strength, and high heat resistance.
Catalog No. 50-276-7800 Supplier Grainger Supplier No. 16Z146
The Silicone Round Cord Metal is a silicone rubber cord with a metal core. It is made of high-quality silicone rubber that is flexible and durable. The metal core provides strength and shape retention. This cord has a round cross-section and comes in multiple diameters from 1/8" to 3/4". It can withstand temperatures from -80°F to 500°F. Potential applications include gaskets, seals, vibration dampening, noise reduction, and more. This product offers flexibility, strength, and high heat resistance.