Learn More
Grainger K4796 Silicone Round Cord 3/8 D 100 L 70A Blue
Supplier:  Grainger 16Z168
Encompass_Preferred
The Silicone Round Cord Metal is a silicone rubber cord with a metal core that is flexible and durable. It has a temperature range of -60°F to 500°F and is resistant to weather, ozone, sunlight, and aging. The cord has a round cross-section and comes in multiple diameters and lengths. It can be used for a variety of applications including weatherstripping, sealing, binding, and more. The metal core provides strength while the silicone jacket offers flexibility. This product is made in the USA.
Catalog No. 50-276-8026