Learn More
Grainger K4796 Silicone Round Cord 5/16 D 50 L 70A Blue
Catalog No. 502767914
Encompass_Preferred
The Silicone Round Cord from Grainger is a flexible, durable silicone cord that can withstand temperatures from -100 to 500°F. It has a round cross-section and is available in multiple diameters from 1/16 inch to 3/4 inch. The cord comes in standard lengths of 25 feet and 50 feet and has excellent resistance to weather, moisture, and UV rays. This silicone cord can be used for a variety of applications including weatherstripping, gasketing, binding, and tying. It will not melt or burn and provides a long service life. The cord has good chemical resistance and will not mark or stain surfaces.
Catalog No. 50-276-7914 Supplier Grainger Supplier No. 16Z163
The Silicone Round Cord from Grainger is a flexible, durable silicone cord that can withstand temperatures from -100 to 500°F. It has a round cross-section and is available in multiple diameters from 1/16 inch to 3/4 inch. The cord comes in standard lengths of 25 feet and 50 feet and has excellent resistance to weather, moisture, and UV rays. This silicone cord can be used for a variety of applications including weatherstripping, gasketing, binding, and tying. It will not melt or burn and provides a long service life. The cord has good chemical resistance and will not mark or stain surfaces.