Learn More
Grainger K4796 Silicone Round Cord 5/16 D 25 L 70A Blue
Catalog No. 502768185
Encompass_Preferred
The Silicone Round Cord Metal is a silicone rubber cord with a metal core that is flexible and durable. It has a temperature range of -60°F to 500°F and comes in multiple diameters and lengths. The cord has good electrical insulation properties and high tensile strength. It is often used for applications like weatherstripping, gasketing, binding, and holding. This product can provide flexibility while maintaining shape in high heat environments. The metal core offers rigidity while the silicone exterior provides insulation against electricity, moisture, and weather.
Catalog No. 50-276-8185 Supplier Grainger Supplier No. 16Z162
The Silicone Round Cord Metal is a silicone rubber cord with a metal core that is flexible and durable. It has a temperature range of -60°F to 500°F and comes in multiple diameters and lengths. The cord has good electrical insulation properties and high tensile strength. It is often used for applications like weatherstripping, gasketing, binding, and holding. This product can provide flexibility while maintaining shape in high heat environments. The metal core offers rigidity while the silicone exterior provides insulation against electricity, moisture, and weather.