Learn More
Grainger K4796 Silicone Round Cord 5/8 D 50 L 70A Blue
Supplier:  Grainger 16Z179
Encompass_Preferred
The Silicone Round Cord from Grainger is a flexible, durable silicone cord that can withstand temperatures from -100 to 500°F. It has a round cross-section and comes in multiple diameters from 1/16 inch to 3/4 inch. The cord is made from platinum-cured silicone which provides excellent resistance to weathering, aging, moisture, and UV rays. With its high flexibility and temperature resistance, this silicone cord is ideal for applications like gaskets, seals, hose assemblies, and more. The cord comes in standard 100 foot spools.
Catalog No. 50-276-7875