Learn More
Grainger K4796 Silicone RoundCord 3/16 D 100 L 70A Blue
Catalog No. 502768077
Encompass_Preferred
The Fisherbrand Silicone Roundcord 3/16" is a flexible, durable, and heat-resistant silicone tubing with a 3/16" diameter. It has a temperature range of -100°C to 204°C and is made of platinum-cured silicone. With a durometer hardness of 50A and tensile strength of 1025 psi, it is sold by the foot in various colors. Latex-free and non-toxic, it suits pharmaceutical, food, beverage, and medical uses. It withstands sterilization, resists oxidation and many chemicals, and provides a tight seal for gasketing, sealing, insulating, protecting, and cushioning.
Catalog No. 50-276-8077 Supplier Grainger Supplier No. 16Z156
The Fisherbrand Silicone Roundcord 3/16" is a flexible, durable, and heat-resistant silicone tubing with a 3/16" diameter. It has a temperature range of -100°C to 204°C and is made of platinum-cured silicone. With a durometer hardness of 50A and tensile strength of 1025 psi, it is sold by the foot in various colors. Latex-free and non-toxic, it suits pharmaceutical, food, beverage, and medical uses. It withstands sterilization, resists oxidation and many chemicals, and provides a tight seal for gasketing, sealing, insulating, protecting, and cushioning.