Learn More
Grainger K4796 Silicone RoundCord 3/32 D 100 L 70A Blue
Catalog No. 502767217
Encompass_Preferred
The Fisherbrand Silicone Roundcord 3/16" is a 3/16" diameter silicone tubing that is flexible, durable, and heat resistant. It is made of platinum-cured silicone and has operating temperature range of -100°C to 260°C. This clear tubing has inside diameter of 0.187" and outside diameter of 0.3125". It meets USP Class VI standards and is non-toxic, non-hemolytic, and non-pyrogenic. The silicone tubing is sold by the foot, with standard lengths starting at 1 foot. It can be used for pharmaceutical, biotechnology, food, beverage, and other applications requiring flexible, chemically inert tubing.
Catalog No. 50-276-7217 Supplier Grainger Supplier No. 16Z144
The Fisherbrand Silicone Roundcord 3/16" is a 3/16" diameter silicone tubing that is flexible, durable, and heat resistant. It is made of platinum-cured silicone and has operating temperature range of -100°C to 260°C. This clear tubing has inside diameter of 0.187" and outside diameter of 0.3125". It meets USP Class VI standards and is non-toxic, non-hemolytic, and non-pyrogenic. The silicone tubing is sold by the foot, with standard lengths starting at 1 foot. It can be used for pharmaceutical, biotechnology, food, beverage, and other applications requiring flexible, chemically inert tubing.