Learn More
Grainger K4796 Silicone RoundCord 5/16 D 100 L 70A Blue
Catalog No. 502768076
Encompass_Preferred
The Silicone Round Cord from Grainger is a flexible, durable silicone cord that can withstand temperatures from -100 to 500°F. It has a round cross-section and comes in multiple diameters from 1/16 inch to 3/4 inch. The cord is made from high quality platinum-cured silicone that resists weathering and aging. It can be used for a variety of applications including binding, strapping, and bundling. The cord has good electrical insulation properties. It comes in standard lengths of 25 ft and 50 ft. This silicone cord offers flexibility, strength, and durability.
Catalog No. 50-276-8076 Supplier Grainger Supplier No. 16Z164
The Silicone Round Cord from Grainger is a flexible, durable silicone cord that can withstand temperatures from -100 to 500°F. It has a round cross-section and comes in multiple diameters from 1/16 inch to 3/4 inch. The cord is made from high quality platinum-cured silicone that resists weathering and aging. It can be used for a variety of applications including binding, strapping, and bundling. The cord has good electrical insulation properties. It comes in standard lengths of 25 ft and 50 ft. This silicone cord offers flexibility, strength, and durability.