Learn More
LifeSensors K48-Linked Tetra-Ubiquitin (Ub4) 25ug
SDP
Supplier:  LifeSensors SI48040025
Encompass_Preferred
Poly-ubiquitylation of target proteins through linkage at K48 is the most thoroughly studied of the various ubiquitin chain linkages. It is now clear that many, if not all, poly-Ub chain topologies are likely to play distinct and important roles in regulating cellular processes. Nevertheless, K48 linkages remain a critical pathway for cells to maintain protein homeostasis and remain intriguing for the study of DUBs that play a role in regulating degradation, as well as the proteasome itself. These tetra-ubiquitin chains generated from the enzymatic linkage (E2-25K) of wild-type ubiquitin through lysine 48. The most distal ubiquitin contains an arginine substitution for the lysine at position 48, limiting chain length.
Catalog No. NC2144452