Learn More
LifeSensors K48-Linked Di-Ubiquitin (Ub2) 100µg
SDP
Supplier:  LifeSensors SI48020100
Encompass_Preferred
Poly-ubiquitylation of target proteins through linkage at K48, is now the most thoroughly studied of the various chain linkages, and we once considered the hallmark of this post-translational modification. K48 di-ubiquitin or K48 linked di-ubiquitin was at the forefront of this research. It is now clear that many, if not all, poly-Ub chain topologies likely play distinct and important roles in regulating cellular processes. Nevertheless, K48 linkage remains a critical pathway for the cells to maintain homeostasis through proteolytic degradation, and as such remains very intriguing for the study of DUBs that play a role in the degradation, as well as the proteasome itself. These di-ubiquitin chains generated from the enzymatic linkage of wild-type ubiquitin through lysine 48. The most distal ubiquitin contains an arginine substitution for the lysine at position 48, limiting chain length.
Catalog No. 50-103-7637