Learn More
LifeSensors Tandem Ubiquitin Binding Entities (K48 HF TUBE) Magnetic 1mL
SDP
Catalog No. NC1726534
Encompass_Preferred
The LifeSensors K48 TUBE HF Magnetic Beads are highly effective for isolating K48-linked polyubiquitinated proteins. These beads are designed for scientific applications and allow for a convenient and efficient one-step pull-down method. They offer protection against degradation and enhance the analysis of polyubiquitinated proteins, making them a valuable resource for western blotting and proteomic studies.
  • High selectivity for K48-linked polyubiquitin chains
  • Affinity of approximately 20 nM for target linkages
  • Streamlined magnetic bead format enables efficient isolation
  • Protects proteins from degradation by deubiquitinating enzymes
  • Eliminates need for epitope-tagged ubiquitin overexpression
Catalog No. NC1726534 Supplier LifeSensors Supplier No. UM0407M1000
The LifeSensors K48 TUBE HF Magnetic Beads are highly effective for isolating K48-linked polyubiquitinated proteins. These beads are designed for scientific applications and allow for a convenient and efficient one-step pull-down method. They offer protection against degradation and enhance the analysis of polyubiquitinated proteins, making them a valuable resource for western blotting and proteomic studies.
  • High selectivity for K48-linked polyubiquitin chains
  • Affinity of approximately 20 nM for target linkages
  • Streamlined magnetic bead format enables efficient isolation
  • Protects proteins from degradation by deubiquitinating enzymes
  • Eliminates need for epitope-tagged ubiquitin overexpression